cis-Cyclooctene 95% stabilised
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(Z)-Cyclooctene
A13477-AD
A13477-AK
A13477-AE
A13477-AP
CAAAAA13477-ADEA
71.57
CAD
CAAAAA13477-AD
CAAAAA13477-AK
CAAAA13477-AE
CAAAA13477-AP
cis-Cyclooctene 95% stabilised
cis-Cyclooctene
Purity: 95%
CAS No.: 931-87-3
Molecular Formula: C8H14
Formula Weight: 110.20
Flash Point: 25°(77°F)
Refractive Index: 1.4700
Odor: Recognizable
Melting Point: -16°
Boiling Point: 145-146°
Beilstein Reference: 1280166
UN No.: 3295
MDL No.: MFCD00001753
Formula:
C₈H₁₄ Boiling Pt: 145…146 °C Melting Pt: –16 °C Density: 0.848 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001753 CAS Number: 931-87-3 EINECS: 213-243-4 UN: 3295 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...