Ethyl indol-3-ylacetate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13474-06
A13474-14
CAAAA13474-06EA
110.13
CAD
CAAAA13474-06
CAAAA13474-14
CAAAA13474-22
Ethyl indol-3-ylacetate 98+%
Ethyl indol-3-ylacetate
Purity: 98+%
CAS No.: 778-82-5
Molecular Formula: C12H13NO2
Formula Weight: 203.24
Flash Point:
Formula:
C₁₂H₁₃NO₂ Boiling Pt: 164…166 °C/1 mm Melting Pt: 41…44 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005635 CAS Number: 778-82-5 EINECS: 212-296-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...