5-Sulfosalicylic acid dihydrate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13461-0B
A13461-30
CAAAA13461-0BEA
237
CAD
CAAAA13461-0B
CAAAA13461-30
5-Sulfosalicylic acid dihydrate 98%
5-Sulphosalicylic acid dihydrate
MDL: MFCD00007508
Beilstein: 650741
Soluble in water
Purity: 98%
CAS No.: 5965-83-3
Molecular Formula: C7H6O6SH.2H2O
Formula Weight: 254.22 (218.19anhy)
Physical Form: Cryst powder
Sensitivity: Light Sensitive
Appearance: White
Odor: Pungent
Melting Point: 103-112°
Solubility: Soluble in water
Beilstein Reference: 650741
Merck Reference: 14,8964
UN No.: 2585
MDL No.: MFCD00007508
Formula:
HO₃SC₆H₃-2-(OH)CO₂H·2H₂O Melting Pt: 103…112 °C Density: 1.695 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007508 CAS Number: 5965-83-3 EINECS: 202-555-6 UN: 2585 ADR: 8,III Merck Index: 14,8964 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...