1-Chloro-4-iodobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A13459-14 A13459-22 A13459-36
CAAAA13459-14EA 70.15 CAD
CAAAA13459-14 CAAAA13459-22 CAAAA13459-36
1-Chloro-4-iodobenzene 99%
1-Chloro-4-iodobenzene

Purity: 99% CAS No.: 637-87-6 Molecular Formula: C6H4ClI Formula Weight: 238.46 Flash Point: 108°(226°F) Melting Point: 52-56° Boiling Point: 226-227° Beilstein Reference: 1634414 MDL No.: MFCD00001053

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 226…227 °C
Melting Pt: 52…56 °C
Density: 1,886 g/cm³ (57 °C)
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001053
CAS Number: 637-87-6
EINECS: 211-305-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR