5-Fluorouracil 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

A13456-06 A13456-14 A13456-22
CAAAA13456-06EA 153.1 CAD
CAAAA13456-06 CAAAA13456-14 CAAAA13456-22
5-Fluorouracil 99%
5-Fluorouracil

Purity: 99% CAS No.: 51-21-8 Molecular Formula: C4H3FN2O2 Formula Weight: 130.08 Melting Point: ca 282° dec. Beilstein Reference: 127172 Merck Reference: 14,4181 UN No.: 2811 MDL No.: MFCD00006018

Formula: C₄H₃FN₂O₂
MW: 130.08 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: ∼ 282 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006018
CAS Number: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,4181

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR