4-Iodophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13455-06
A13455-14
A13455-22
CAAAAA13455-06EA
42.66
CAD
CAAAAA13455-06
CAAAA13455-14
CAAAA13455-22
CAAAA13455-36
4-Iodophenol 98+%
4-Iodophenol
Purity: 98+%
CAS No.: 540-38-5
Molecular Formula: C6H5IO
Formula Weight: 220.01
Melting Point: 91-95°
Boiling Point: 138°/5mm
Merck Reference: 14,5036
MDL No.: MFCD00002327
Formula:
C₆H₅IO Boiling Pt: 138 °C/5 mm Melting Pt: 91…95 °C Density: 2.13 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002327 CAS Number: 540-38-5 EINECS: 208-745-5 UN: 3261 ADR: 8,III Merck Index: 14,5036 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...