1,3,5-Triisopropylbenzene 95%
Supplier: Thermo Scientific Chemicals
Warning
|
A13453-0B
A13453-18
A13453-30
CAAAA13453-0BEA
1145.5
CAD
CAAAA13453-0B
CAAAA13453-18
CAAAA13453-30
1,3,5-Triisopropylbenzene 95%
1,3,5-Triisopropylbenzene
Purity: 95%
CAS No.: 717-74-8
Molecular Formula: C15H24
Formula Weight: 204.36
Flash Point: 86°(186°F)
Refractive Index: 1.4880
Melting Point: -7°
Boiling Point: 235-237°
Beilstein Reference: 1862750
MDL No.: MFCD00008890
Formula:
C₁₅H₂₄ MW: 204.36 g/mol Boiling Pt: 232 Melting Pt: -7 Density: 0.854 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008890 CAS Number: 717-74-8 EINECS: 211-941-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...