Sodium octanoate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate

A13451-22 A13451-36
CAAAA13451-22EA 92.9 CAD
CAAAA13451-22 CAAAA13451-36
Sodium octanoate 96%
Sodium octanoate

Purity: 96% CAS No.: 1984-06-1 Molecular Formula: CH3(CH2)6CO2Na Formula Weight: 166.20 Melting Point: 300° Beilstein Reference: 3597723

Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
Melting Pt: >300 °C
Density: Min. 0.9949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR