α-Chloro-o-xylene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13448-22
A13448-36
CAAAA13448-22EA
136.04
CAD
CAAAA13448-22
CAAAA13448-36
α-Chloro-o-xylene 98+%
α-Chloro-o-xylene
Purity: 98+%
CAS No.: 552-45-4
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Storage Temperature: 2°C to 8°C
Flash Point: 73°(163°F)
Refractive Index: 1.5410
Odor: Pungent
Melting Point: 2°
Boiling Point: 197-199°
Beilstein Reference: 386016
Merck Reference: 14,10091
UN No.: 3265
MDL No.: MFCD00000899
Formula:
C₈H₉Cl Boiling Pt: 197…199 °C Melting Pt: 2 °C Density: 1.081 Flash Pt: 73 °C (163 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000899 CAS Number: 552-45-4 EINECS: 209-013-8 UN: 3265 ADR: 8,III Merck Index: 14,10091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...