2-Iodo-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13439-06
A13439-14
CAAAAA13439-22EA
1635.96
CAD
CAAAAA13439-22
CAAAA13439-06
CAAAA13439-14
2-Iodo-m-xylene 98%
2-Iodo-m-xylene
Purity: 98%
CAS No.: 608-28-6
Molecular Formula: C8H9I
Formula Weight: 232.07
Flash Point: 110°(230°F)
Refractive Index: 1.6025
Boiling Point: 230-231°
Beilstein Reference: 1857414
MDL No.: MFCD00019012
Formula:
C₈H₉I Boiling Pt: 230…231 °C Melting Pt: 11,2 °C Density: 1,620 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00019012 CAS Number: 608-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...