1,5-Diazobicyclo[4.3.0]-5-nonene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13437-14
A13437-22
CAAAA13437-14EA
118.82
CAD
CAAAA13437-14
CAAAA13437-22
1,5-Diazobicyclo[4.3.0]-5-nonene 98%
1,5-Diazobicyclo[4.3.0]-5-nonene
Purity: 98%
CAS No.: 3001-72-7
Molecular Formula: C7H12N2
Formula Weight: 124.19
Flash Point: 94°(201°F)
Refractive Index: 1.5200
Boiling Point: 95-98°/7mm
Beilstein Reference: 2417
UN No.: 3267
MDL No.: MFCD00005554
Formula:
C₇H₁₂N₂ Boiling Pt: 95…98 °C/7 mm Density: 1.042 Flash Pt: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005554 CAS Number: 3001-72-7 EINECS: 221-087-3 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...