(±)-4-Methyl-2-pentanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Methyl isobutyl carbinol

A13435-0F A13435-AP
CAAAA13435-0FEA 111.55 CAD
CAAAA13435-0F CAAAA13435-AP
(±)-4-Methyl-2-pentanol 99%
(±)-4-Methyl-2-pentanol

Purity: 99% CAS No.: 108-11-2 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 41°(105°F) Refractive Index: 1.4110 Odor: Slightly sweetish Melting Point: -90° Boiling Point: 131-133° Beilstein Reference: 1098268 UN No.: 2053 MDL No.: MFCD00004550

Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 131…133 °C
Melting Pt: –90 °C
Density: 0.806
Flash Pt: 41 °C (105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004550
CAS Number: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR