trans-3-Hexenoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13428-09
A13428-18
CAAAA13428-09EA
70.78
CAD
CAAAA13428-09
CAAAA13428-18
CAAAA13428-30
trans-3-Hexenoic acid 97%
trans-3-Hexenoic acid
Purity: 97%
CAS No.: 1577-18-0
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 99°(210°F)
Refractive Index: 1.4398
Odor: Rancid
Melting Point: 11-12°
Boiling Point: 206°
Beilstein Reference: 1700862
UN No.: 3265
MDL No.: MFCD00002786
Formula:
C₆H₁₀O₂ MW: 114.14 g/mol Boiling Pt: 206 °C Melting Pt: 11…12 °C Density: 0.961 Flash Pt: 99 °C (210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002786 CAS Number: 1577-18-0 EINECS: 216-417-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...