3-Chlorocinnamic acid predominantly trans 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(3-Chlorophenyl)acrylic acid, (2E)-3-(3-Chlorophenyl)acrylic acid

A13416-09 A13416-18 A13416-30
CAAAA13416-09EA 60.36 CAD
CAAAA13416-09 CAAAA13416-18 CAAAA13416-30
3-Chlorocinnamic acid predominantly trans 98+%
3-Chlorocinnamic acid

Purity: 98+% CAS No.: 1866-38-2 Molecular Formula: C9H7ClO2 Formula Weight: 182.61 Melting Point: 161-164° Beilstein Reference: 2207378

Formula: C₉H₇ClO₂
Melting Pt: 161…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00004384
CAS Number: 1866-38-2
EINECS: 217-478-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR