(2-Nitrophenyl)acetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13411-14
A13411-22
CAAAA13411-14EA
74.1
CAD
CAAAA13411-14
CAAAA13411-22
CAAAA13411-36
(2-Nitrophenyl)acetic acid 99%
(2-Nitrophenyl)acetic acid
Purity: 99%
CAS No.: 3740-52-1
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Odor: Odorless
Melting Point: 139-142°
Beilstein Reference: 1959243
MDL No.: MFCD00007190
Formula:
(C₆H₄NO₂)CH₂COOH Melting Pt: 139…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007190 CAS Number: 3740-52-1 EINECS: 223-128-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...