2,5-Dibromothiophene 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13406-14
A13406-18
A13406-30
CAAAAA13406-14EA
73.31
CAD
CAAAAA13406-14
CAAAA13406-18
CAAAA13406-30
2,5-Dibromothiophene 96%
2,5-Dibromothiophene
Purity: 96%
CAS No.: 3141-27-3
Molecular Formula: C4H2Br2S
Formula Weight: 241.94
Flash Point: 99°(210°F)
Refractive Index: 1.6290
Odor: Pungent
Melting Point: -6°
Boiling Point: 210-212°
Beilstein Reference: 109899
UN No.: 2810
MDL No.: MFCD00005420
Formula:
C₄H₂Br₂S Boiling Pt: 210…212 °C Melting Pt: –6 °C Density: 2.150 Flash Pt: 99 °C(210 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005420 CAS Number: 3141-27-3 EINECS: 221-547-3 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...