Pyrogallol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13405-22
A13405-36
CAAAA13405-22EA
108.07
CAD
CAAAA13405-22
CAAAA13405-36
Pyrogallol 98+%
Pyrogallol
Purity: 98+%
CAS No.: 87-66-1
Molecular Formula: C6H6O3
Formula Weight: 126.11
Odor: Odorless
Melting Point: 131-135°
Boiling Point: 309°
Beilstein Reference: 907431
Merck Reference: 14,8000
UN No.: 2811
MDL No.: MFCD00002192
Formula:
C₆H₆O₃ Boiling Pt: 309 °C Melting Pt: 131…135 °C Density: 1.45 Storage Temperature: Ambient |
MDL Number:
MFCD00002192 CAS Number: 87-66-1 EINECS: 201-762-9 UN: 2811 ADR: 6.1,III Merck Index: 14,8000 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...