2,4-Difluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A13403-06 A13403-14
CAAAAA13403-22EA 686.81 CAD
CAAAAA13403-22 CAAAA13403-06 CAAAA13403-14
2,4-Difluorobenzoic acid 98%
2,4-Difluorobenzoic acid

Purity: 98% CAS No.: 1583-58-0 Molecular Formula: C7H4F2O2 Formula Weight: 158.11 Odor: Odorless Melting Point: 186-190° Beilstein Reference: 973355 MDL No.: MFCD00011670

Formula: C₇H₄F₂O₂
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00011670
CAS Number: 1583-58-0
EINECS: 216-430-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR