Thiosalicylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13401-0B
A13401-18
A13401-30
CAAAAA13401-0BEA
1252.94
CAD
CAAAAA13401-0B
CAAAA13401-18
CAAAA13401-30
Thiosalicylic acid 98%
Thiosalicylic acid
Purity: 98%
CAS No.: 147-93-3
Molecular Formula: C7H6O2S
Formula Weight: 154.19
Odor: Stench
Melting Point: 165-168°
Beilstein Reference: 508507
Merck Reference: 14,9360
MDL No.: MFCD00004836
Formula:
C₇H₆O₂S Boiling Pt: 230 °C (0.02 -rr) Melting Pt: 165…168 °C Density: 1.49 Storage Temperature: Ambient |
MDL Number:
MFCD00004836 CAS Number: 147-93-3 EINECS: 205-704-3 UN: 3335 ADR: 9, Merck Index: 14,9360 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...