3,4-Dimethoxybenzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

A13396-14 A13396-22 A13396-36
CAAAA13396-14EA 79.63 CAD
CAAAA13396-14 CAAAA13396-22 CAAAA13396-36
3,4-Dimethoxybenzyl alcohol 97%
3,4-Dimethoxybenzyl alcohol

Purity: 97% CAS No.: 93-03-8 Molecular Formula: C9H12O3 Formula Weight: 168.19 Flash Point: 110°(230°F) Refractive Index: 1.5530 Melting Point: ca 22° Boiling Point: 297-298° Beilstein Reference: 639388 MDL No.: MFCD00004638

Formula: C₉H₁₂O₃
Boiling Pt: 297…298 °C
Melting Pt: ∼ 22 °C
Density: 1.176
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR