3-Methylthiophene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13394-14
A13394-22
CAAAAA13394-14EA
64.15
CAD
CAAAAA13394-14
CAAAAA13394-22
CAAAA13394-36
3-Methylthiophene 98+%
3-Methylthiophene
Purity: 98+%
CAS No.: 616-44-4
Molecular Formula: C5H6S
Formula Weight: 98.17
Flash Point: 11°(51°F)
Refractive Index: 1.5200
Odor: Aromatic
Melting Point: -69°
Boiling Point: 115-117°
Beilstein Reference: 1300
UN No.: 1993
MDL No.: MFCD00005470
Formula:
C₅H₆S Boiling Pt: 115…117 °C Melting Pt: –69 °C Density: 1.020 Flash Pt: 11 °C (51 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005470 CAS Number: 616-44-4 EINECS: 210-482-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...