(±)-4-Fluoromandelic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13391-03
A13391-06
A13391-14
CAAAA13391-03EA
27.97
CAD
CAAAA13391-03
CAAAA13391-06
CAAAA13391-14
(±)-4-Fluoromandelic acid 98%
(±)-4-Fluoromandelic acid
Purity: 98%
CAS No.: 395-33-5
Molecular Formula: C8H7FO3
Formula Weight: 170.14
Melting Point: 133-138°
Beilstein Reference: 2211001
Formula:
C₈H₇FO₃ Melting Pt: 133…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044402 CAS Number: 395-33-5 EINECS: 206-899-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...