Diethanolamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ß,ß′-Dihydroxydiethylamine, Bis-(ß-hydroxyethyl)-amine, 2,2'-Iminodiethanol

A13389-30 A13389-0E A13389-36
CAAAAA13389-30EA 39.82 CAD
CAAAAA13389-30 CAAAA13389-0E CAAAA13389-36
Diethanolamine 99%
Diethanolamine

Purity: 99% CAS No.: 111-42-2 Molecular Formula: C4H11NO2 Formula Weight: 105.14 Flash Point: 176°(349°F) Physical Form: Low-melting solid Density: 1.097 Refractive Index: 1.4770 Sensitivity: Hygroscopic Appearance: White Odor: Ammonia-like pH: 11 Melting Point: 27-30° Boiling Point: 268-270° Beilstein Reference: 605315 Merck Reference: 14,3107 MDL No.: MFCD00002843

Formula: C₄H₁₁NO₂
MW: 105.14 g/mol
Boiling Pt: 268…270 °C
Melting Pt: 27…30 °C
Density: 1.097
Flash Pt: 176 °C (349 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
UN: 3259
ADR: 8,III
REACH: 01-2119488930-28
Merck Index: 14,3107

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR