4,5-Dibromo-2-thiophenesulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13384-14
A13384-03
A13384-06
CAAAAA13384-14EA
1049.12
CAD
CAAAAA13384-14
CAAAA13384-03
CAAAA13384-06
4,5-Dibromo-2-thiophenesulfonyl chloride 97%
4,5-Dibromo-2-thiophenesulphonyl chloride
Purity: 97%
CAS No.: 81606-31-7
Molecular Formula: C4HBr2ClO2S2
Formula Weight: 340.45
Melting Point: 75-79°
Beilstein Reference: 162960
UN No.: 3261
MDL No.: MFCD00041439
Formula:
C₄HBr₂ClO₂S₂ Melting Pt: 75…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041439 CAS Number: 81606-31-7 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...