Methanesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13383-22
A13383-36
CAAAA13383-22EA
42.03
CAD
CAAAA13383-22
CAAAA13383-36
Methanesulfonyl chloride 98%
Methanesulphonyl chloride
Purity: 98%
CAS No.: 124-63-0
Molecular Formula: CH3ClO2S
Formula Weight: 114.55
Flash Point: 97°(206°F)
Refractive Index: 1.4520
Odor: Pungent
Melting Point: -33°
Boiling Point: 161°
Beilstein Reference: 506297
Merck Reference: 14,5955
UN No.: 3246
MDL No.: MFCD00007454
Formula:
CH₃ClO₂S Boiling Pt: 161 °C Melting Pt: –33 °C Density: 1.477 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007454 CAS Number: 124-63-0 EINECS: 204-706-1 UN: 3246 ADR: 6.1,I Merck Index: 14,5955 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...