2-Mercaptopyrimidine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Thiopyrimidine

A13382-22 A13382-09 A13382-30
CAAAAA13382-22EA 467.68 CAD
CAAAAA13382-22 CAAAA13382-09 CAAAA13382-30
2-Mercaptopyrimidine 98%
2-Mercaptopyrimidine

Purity: 98% CAS No.: 1450-85-7 Molecular Formula: C4H4N2S Formula Weight: 112.15 Melting Point: ca 230° dec. Beilstein Reference: 107105

Formula: C₄H₄N₂S
MW: 112.16 g/mol
Melting Pt: ∼ 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006073
CAS Number: 1450-85-7
EINECS: 215-917-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR