1,3-Dimethoxybenzene 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Resorcinol dimethyl ether
A13380-22
A13380-36
CAAAA13380-22EA
53.09
CAD
CAAAA13380-22
CAAAA13380-36
1,3-Dimethoxybenzene 98%
1,3-Dimethoxybenzene
Purity: 98%
CAS No.: 151-10-0
Molecular Formula: C8H10O2
Formula Weight: 138.17
Flash Point: 87°(188°F)
Refractive Index: 1.5250
Odor: Characteristic
Melting Point: -52°
Boiling Point: 217-218°
Beilstein Reference: 878582
MDL No.: MFCD00008384
Formula:
C₈H₁₀O₂ Boiling Pt: 217…218 °C Melting Pt: –52 °C Density: 1.065 Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008384 CAS Number: 151-10-0 EINECS: 205-783-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...