α,α,α,4-Tetrafluorotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluorobenzotrifluoride, 1-Fluoro-4-(trifluoromethyl)benzene

A13377-06 A13377-14
CAAAA13377-06EA 75.84 CAD
CAAAA13377-06 CAAAA13377-14
α,α,α,4-Tetrafluorotoluene 98+%
α,α,α,4-Tetrafluorotoluene

Purity: 98+% CAS No.: 402-44-8 Molecular Formula: C7H4F4 Formula Weight: 164.10 Flash Point: 10°(50°F) Refractive Index: 1.4010 Melting Point: -42° Boiling Point: 103-105° Beilstein Reference: 1909390 UN No.: 1993 MDL No.: MFCD00000399

Formula: C₇H₄F₄
MW: 164.1 g/mol
Boiling Pt: 103…105 °C
Melting Pt: –42 °C
Density: 1.295
Flash Pt: 10 °C (50 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000399
CAS Number: 402-44-8
EINECS: 206-944-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR