2-Butylfuran 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13376-09
A13376-18
A13376-30
CAAAA13376-09EA
84.06
CAD
CAAAA13376-09
CAAAA13376-18
CAAAA13376-30
2-Butylfuran 98%
2-Butylfuran
Purity: 98%
CAS No.: 4466-24-4
Molecular Formula: C8H12O
Formula Weight: 124.18
Storage Temperature: 2°C to 8°C
Flash Point: 32°(89°F)
Odor: Pleasant
Boiling Point: 153-154°
Beilstein Reference: 107045
UN No.: 1993
MDL No.: MFCD00047071
Formula:
C₈H₁₂O Boiling Pt: 153…154 °C Density: 0.886 Flash Pt: 32 °C(89 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00047071 CAS Number: 4466-24-4 EINECS: 224-732-7 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...