4-Bromocumene 97%

Supplier: Thermo Scientific Chemicals
Warning

A13370-09 A13370-06
CAAAAA13370-09EA 211.72 CAD
CAAAAA13370-09 CAAAAA13370-18 CAAAA13370-06
4-Bromocumene 97%
4-Bromocumene

MDL: MFCD00039159 Beilstein: 1858096 Insoluble in water


Purity: 97% CAS No.: 586-61-8 Molecular Formula: C9H11Br Formula Weight: 199.10 Flash Point: 100°(212°F) Refractive Index: 1.5360 Melting Point: -12° Boiling Point: 219° Solubility: Insoluble in water Beilstein Reference: 1858096 UN No.: 3082 MDL No.: MFCD00039159

Formula: C₉H₁₁Br
Boiling Pt: 219 °C
Melting Pt: –12 °C
Density: 1.315
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039159
CAS Number: 586-61-8
EINECS: 209-577-5
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR