ω-Bromo-3,4-dichloroacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13365-06
A13365-14
A13365-22
CAAAA13365-06EA
88.32
CAD
CAAAA13365-06
CAAAA13365-14
CAAAA13365-22
ω-Bromo-3,4-dichloroacetophenone 98%
ω-Bromo-3,4-dichloroacetophenone
Purity: 98%
CAS No.: 2632-10-2
Molecular Formula: C8H5BrCl2O
Formula Weight: 267.94
Storage Temperature: 2°C to 8°C
Melting Point: 55-58°
Beilstein Reference: 511981
UN No.: 1759
MDL No.: MFCD00051581
Formula:
C₈H₅BrCl₂O Melting Pt: 55…58 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051581 CAS Number: 2632-10-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...