Pyrazine-2-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
A13363-14
A13363-22
CAAAA13363-14EA
67.15
CAD
CAAAA13363-14
CAAAA13363-22
CAAAA13363-36
Pyrazine-2-carboxylic acid 99%
Pyrazine-2-carboxylic acid
Purity: 99%
CAS No.: 98-97-5
Molecular Formula: C5H4N2O2
Formula Weight: 124.10
Melting Point: ca 225° dec.
Beilstein Reference: 112305
Merck Reference: 14,7959
MDL No.: MFCD00006130
Formula:
C₅H₄N₂O₂ Boiling Pt: 313 °C (1013 hPa) Melting Pt: ∼ 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006130 CAS Number: 98-97-5 EINECS: 202-718-1 Merck Index: 14,7959 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...