4-Tolylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13347-03
A13347-06
A13347-14
CAAAA13347-03EA
67.31
CAD
CAAAA13347-03
CAAAA13347-06
CAAAA13347-14
4-Tolylboronic acid 98%
4-Tolylboronic acid
Purity: 98%
CAS No.: 5720-05-8
Molecular Formula: C7H9BO2
Formula Weight: 135.96
Melting Point: 246-254°
Beilstein Reference: 2935970
Formula:
C₇H₉BO₂ Melting Pt: 246…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039138 CAS Number: 5720-05-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...