1,4-Diethylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13343-06
A13343-14
A13343-22
CAAAA13343-06EA
92.75
CAD
CAAAA13343-06
CAAAA13343-14
CAAAA13343-22
1,4-Diethylbenzene 98%
1,4-Diethylbenzene
Purity: 98%
CAS No.: 105-05-5
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 55°(131°F)
Refractive Index: 1.4950
Odor: Aromatic
Melting Point: -43°
Boiling Point: 182-184°
Beilstein Reference: 1903396
UN No.: 2049
MDL No.: MFCD00009264
Formula:
C₁₀H₁₄ Boiling Pt: 182…184 °C Melting Pt: –43 °C Density: 0.862 Flash Pt: 55 °C (131 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009264 CAS Number: 105-05-5 EINECS: 203-265-2 UN: 2049 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...