2,4-Dichlorobenzyl alcohol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13338-14
A13338-22
A13338-36
CAAAA13338-14EA
105.07
CAD
CAAAA13338-14
CAAAA13338-22
CAAAA13338-36
2,4-Dichlorobenzyl alcohol 99%
2,4-Dichlorobenzyl alcohol
Purity: 99%
CAS No.: 1777-82-8
Molecular Formula: C7H6Cl2O
Formula Weight: 177.03
Flash Point:
Formula:
C₇H₆Cl₂O Boiling Pt: 150 °C (25 -rr) Melting Pt: 57…61 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004606 CAS Number: 1777-82-8 EINECS: 217-210-5 Merck Index: 14,3060 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...