4-Methylcinnamic acid predominantly trans 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(p-Tolyl)acrylic acid
A13337-06
A13337-14
A13337-22
CAAAA13337-06EA
48.03
CAD
CAAAA13337-06
CAAAA13337-14
CAAAA13337-22
4-Methylcinnamic acid predominantly trans 99%
4-Methylcinnamic acid
Purity: 99%
CAS No.: 1866-39-3
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 196-200°
Beilstein Reference: 1364327
Formula:
C₁₀H₁₀O₂ MW: 162.19 g/mol Boiling Pt: 303 Melting Pt: 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002697 CAS Number: 1866-39-3 EINECS: 217-479-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...