3-Nitrophenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13336-03
A13336-06
A13336-14
CAAAA13336-03EA
39.18
CAD
CAAAA13336-03
CAAAA13336-06
CAAAA13336-14
3-Nitrophenylboronic acid 98%
3-Nitrophenylboronic acid
Purity: 98%
CAS No.: 13331-27-6
Molecular Formula: C6H6BNO4
Formula Weight: 166.93
Melting Point: ca 285° dec.
Beilstein Reference: 2938638
Formula:
C₆H₆BNO₄ Melting Pt: ∼ 285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00007193 CAS Number: 13331-27-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...