6-Methyl-5-hepten-2-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13335-14
A13335-22
CAAAAA13335-14EA
41.87
CAD
CAAAAA13335-14
CAAAA13335-22
6-Methyl-5-hepten-2-one 98%
6-Methyl-5-hepten-2-one
Purity: 98%
CAS No.: 110-93-0
Molecular Formula: C8H14O
Formula Weight: 126.20
Flash Point: 56°(133°F)
Refractive Index: 1.4390
Melting Point: -67°
Boiling Point: 172-174°
Beilstein Reference: 1741705
UN No.: 1224
MDL No.: MFCD00008905
Formula:
C₈H₁₄O Boiling Pt: 172…174 °C Melting Pt: –67 °C Density: 0.855 Flash Pt: 56 °C (133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008905 CAS Number: 110-93-0 EINECS: 203-816-7 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...