2-Amino-5-methylpyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline
A13330-18
A13330-30
CAAAA13330-18EA
128.93
CAD
CAAAA13330-18
CAAAA13330-30
2-Amino-5-methylpyridine 99%
2-Amino-5-methylpyridine
Purity: 99%
CAS No.: 1603-41-4
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 118°(244°F)
Odor: Unpleasant
Boiling Point: 226-228°
Beilstein Reference: 107050
UN No.: 2811
MDL No.: MFCD00006328
Formula:
C₆H₈N₂ Boiling Pt: 226…228 °C Melting Pt: 74…78 °C Flash Pt: 118 °C (244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006328 CAS Number: 1603-41-4 EINECS: 216-503-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...