Methanesulfonic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13328-14
A13328-09
A13328-18
A13328-30
CAAAAA13328-14EA
130.98
CAD
CAAAAA13328-14
CAAAA13328-09
CAAAA13328-18
CAAAA13328-30
Methanesulfonic anhydride 98%
Methanesulphonic anhydride
Purity: 98%
CAS No.: 7143-01-3
Molecular Formula: C2H6O5S2
Formula Weight: 174.20
Melting Point: 65-69°
Boiling Point: 125°/4mm
UN No.: 3261
MDL No.: MFCD00007556
Formula:
C₂H₆O₅S₂ MW: 174.2 g/mol Boiling Pt: 125 °C/4 mm Melting Pt: 65…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007556 CAS Number: 7143-01-3 EINECS: 230-442-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...