N,N-Dimethyl-m-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13326-14
A13326-22
CAAAA13326-14EA
87.53
CAD
CAAAA13326-14
CAAAA13326-22
N,N-Dimethyl-m-toluidine 98%
N,N-Dimethyl-m-toluidine
Purity: 98%
CAS No.: 121-72-2
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 85°(185°F)
Refractive Index: 1.5500
Melting Point: -15°
Boiling Point: 212-215°
Beilstein Reference: 1422766
UN No.: 1708
MDL No.: MFCD00008305
Formula:
C₉H₁₃N Boiling Pt: 212…215 °C Melting Pt: –15 °C Density: 0.932 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008305 CAS Number: 121-72-2 EINECS: 204-495-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...