3-Bromo-4-fluorobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

A13318-06 A13318-14
CAAAA13318-06EA 42.82 CAD
CAAAA13318-06 CAAAA13318-14 CAAAA13318-22
3-Bromo-4-fluorobenzonitrile 98%
3-Bromo-4-fluorobenzonitrile

Purity: 98% CAS No.: 79630-23-2 Molecular Formula: C7H3BrFN Formula Weight: 200.01 Melting Point: 54-58° Boiling Point: 134-136°/33mm UN No.: 3439 MDL No.: MFCD00055432

Formula: C₇H₃BrFN
MW: 200.01 g/mol
Boiling Pt: 134…136 °C/33 mm
Melting Pt: 54…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00055432
CAS Number: 79630-23-2
EINECS: 279-200-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR