2,3,5,6-Tetramethylbenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3,5,6-Tetramethylbenzene-1-sulfonyl chloride, 2,3,5,6-Tetramethylbenzene-1-sulphonyl chloride

A13313-09 A13313-18
CAAAA13313-09EA 72.68 CAD
CAAAA13313-09 CAAAA13313-18 CAAAA13313-30
2,3,5,6-Tetramethylbenzenesulfonyl chloride 98%
2,3,5,6-Tetramethylbenzenesulphonyl chloride

Purity: 98% CAS No.: 60706-63-0 Molecular Formula: C10H13ClO2S Formula Weight: 232.73 Melting Point: 97-101° Beilstein Reference: 1876644 UN No.: 3261 MDL No.: MFCD00014723

Formula: C₁₀H₁₃ClO₂S
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00014723
CAS Number: 60706-63-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR