α-Bromo-p-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13297-06
A13297-14
A13297-22
CAAAA13297-06EA
54.04
CAD
CAAAA13297-06
CAAAA13297-14
CAAAA13297-22
α-Bromo-p-xylene 98%
α-Bromo-p-xylene
Purity: 98%
CAS No.: 104-81-4
Molecular Formula: C8H9Br
Formula Weight: 185.07
Storage Temperature: 2°C to 8°C
Flash Point: 97°(206°F)
Melting Point: 33-36°
Boiling Point: 218-220°
Beilstein Reference: 507389
Merck Reference: 14,10090
UN No.: 3417
MDL No.: MFCD00000181
Formula:
C₈H₉Br Boiling Pt: 218…220 °C Melting Pt: 33…36 °C Density: 1.324 Flash Pt: 97 °C (206 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000181 CAS Number: 104-81-4 EINECS: 203-240-6 UN: 3417 ADR: 6.1,II Merck Index: 14,10090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...