Lauroyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13294-22
A13294-36
CAAAA13294-22EA
80.42
CAD
CAAAA13294-22
CAAAA13294-36
Lauroyl chloride 98%
Lauroyl chloride
Purity: 98%
CAS No.: 112-16-3
Molecular Formula: C12H23ClO
Formula Weight: 218.77
Flash Point: 140°(284°F)
Refractive Index: 1.4460
Melting Point: -17°
Boiling Point: 130-132°/7mm
Beilstein Reference: 1281201
UN No.: 3265
MDL No.: MFCD00000740
Formula:
C₁₂H₂₃ClO Boiling Pt: 130…132 °C/7 mm Melting Pt: –17 °C Density: 0.922 Flash Pt: 140 °C (284 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000740 CAS Number: 112-16-3 EINECS: 203-941-7 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...