Formic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13285-0C
A13285-0E
A13285-36
CAAAA13285-0CEA
494.54
CAD
CAAAA13285-0C
CAAAA13285-0E
CAAAA13285-36
Formic acid 97%
Formic acid
Purity: 97%
CAS No.: 64-18-6
Molecular Formula: CH2O2
Formula Weight: 46.03
Flash Point: 50°(156°F)
Refractive Index: 1.3710
Melting Point: 7-9°
Boiling Point: 100-101°
Beilstein Reference: 1209246
Merck Reference: 14,4241
UN No.: 1779
MDL No.: MFCD00003297
Formula:
HCO₂H Boiling Pt: 100…101 °C Melting Pt: 7…9 °C Density: 1.220 Flash Pt: 50 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8,II Merck Index: 14,4241 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...