Bromoacetaldehyde diethyl acetal 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13276-0E
A13276-22
A13276-36
CAAAA13276-0EEA
722.06
CAD
CAAAA13276-0E
CAAAA13276-22
CAAAA13276-36
Bromoacetaldehyde diethyl acetal 97%
Bromoacetaldehyde diethyl acetal
Purity: 97%
CAS No.: 2032-35-1
Molecular Formula: C6H13BrO2
Formula Weight: 197.08
Storage Temperature: 2°C to 8°C
Flash Point: 61°(142°F)
Refractive Index: 1.4390
Boiling Point: 66-67°/18mm
Beilstein Reference: 635754
UN No.: 2810
MDL No.: MFCD00000214
Formula:
BrH₂CCH(OCH₂CH₃)₂ Boiling Pt: 66…67 °C/18 mm Density: 1.270 Flash Pt: 61 °C (142 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000214 CAS Number: 2032-35-1 EINECS: 217-989-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...