Bromopentafluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromo-2,3,4,5,6-pentafluorobenzene
A13273-36
A13273-14
A13273-22
CAAAAA13273-36EA
2376.32
CAD
CAAAAA13273-36
CAAAA13273-14
CAAAA13273-22
Bromopentafluorobenzene 99%
Bromopentafluorobenzene
Purity: 99%
CAS No.: 344-04-7
Molecular Formula: C6BrF5
Formula Weight: 246.97
Flash Point: 87°(188°F)
Refractive Index: 1.4500
Melting Point: ca -31°
Boiling Point: 136-137°
Beilstein Reference: 1819390
MDL No.: MFCD00000287
Formula:
C₆BrF₅ Boiling Pt: 136…137 °C Melting Pt: ∼ …31 °C Density: 1.981 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000287 CAS Number: 344-04-7 EINECS: 206-449-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...