trans-2-Hexen-1-ol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
trans-Hex-2-en-1-ol
A13272-14
A13272-36
CAAAA13272-14EA
50.72
CAD
CAAAA13272-14
CAAAA13272-36
trans-2-Hexen-1-ol 97%
trans-2-Hexen-1-ol
Purity: 97%
CAS No.: 928-95-0
Molecular Formula: C6H12O
Formula Weight: 100.16
Flash Point: 54°(129°F)
Refractive Index: 1.4380
Odor: Pleasant
Boiling Point: 158-160°
Beilstein Reference: 1719709
UN No.: 1987
MDL No.: MFCD00002927
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 158…160 °C Melting Pt: 65…67 °C Density: 0.843 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002927 CAS Number: 928-95-0 EINECS: 213-191-2 UN: 1987 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...