Benzyltriethylammonium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl triethylammonium chloride

A13268-22 A13268-36
CAAAA13268-22EA 68.73 CAD
CAAAA13268-22 CAAAA13268-36
Benzyltriethylammonium chloride 99%
Benzyltriethylammonium chloride

Purity: 99% CAS No.: 56-37-1 Molecular Formula: (CH3CH2)3N(CH2C6H5)Cl Formula Weight: 227.78 Melting Point: ca 190° dec. Beilstein Reference: 3574984

Formula: C₁₃H₂₂ClN
MW: 227.78 g/mol
Melting Pt: ∼ 190 °C (dec.)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011824
CAS Number: 56-37-1
EINECS: 200-270-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR